Description
Polyethylene pick-up rod with powerful Alnico magnet sealed in one end
- Alnico-V magnet sealed in one end
- Handy loop for hanging on the other end
- 9.5mm diameter
Specifications
Specifications
| Diameter (English) | 0.37 in. |
| For Use With (Application) | Recovering stir bars from flasks and other vessels |
| For Use With (Equipment) | Stirring bars |
| Includes | Alnico-V magnet at one end and hanging loop at other |
| Length (English) | 12 in. |
| Diameter (Metric) | 9.5 mm |
| Length (Metric) | 30.48 cm |
| Material | Polyethylene |