Learn More
Sampling Systems™ SteriWare™ Single Use Production and Sampling Scoops
BIOPHARMA
Product Code. 17564616 Shop All Sampling Systems Products
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
150 mL
250 mL
500 mL
1000 mL
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Pack of 30
Pack of 40
Product Code. Capacity (Metric) Sterility unitSize
17564616 100 mL Non-sterile Pack of 100
17524626 25 mL Non-sterile Pack of 100
17554626 25 mL Sterile Pack of 100
17584626 50 mL Non-sterile Pack of 100
17594616 100 mL Sterile Pack of 100
17504626 150 mL Non-sterile Pack of 100
17514626 150 mL Sterile Pack of 100
17534626 250 mL Non-sterile Pack of 100
17544626 250 mL Sterile Pack of 100
17564626 500 mL Non-sterile Pack of 40
17574626 500 mL Sterile Pack of 40
17574616 1000 mL Non-sterile Pack of 30
17584616 1000 mL Sterile Pack of 30
13 options
Product Code. 17564616
Brand: Sampling Systems™ 8286A100
Only null left
Add to basket
Add to basket
These SteriWare™ Single Use Production and Sampling powder Scoops are ideal for sampling, production and laboratory work. The complete range is made to the highest standards.
  • SteriWare™ flat bottomed powder scoops are single use, free-standing scoops designed for use in GMP manufacturing areas.
  • They are perfect for sampling and dispensing powders and granules.
  • These powder scoops are manufactured in a cleanroom to the highest standards.
  • Everything from the ergonomic design to the class leading documentation means these are the ideal scoop for use in pharmaceutical and food factories.
  • The robust design and ergonomic handle ensures that they are easy and comfortable to use.
  • All powder scoops are individually packed and are available in both sterile and non-sterile versions.
  • Simply choose your scoop size, unpack and use.
  • SteriWare™ Scoops can be used as volumetric scoops by levelling off the excess powder to give you the stated set volume.
  • They are available in a wide ranges of sizes from 25 mL (0.9 oz.) to 1000 mL (36 oz.).
  • Use straight from the bag.
  • The robust, ergonomic handles has been designed so that they can be used repeatedly.
  • If measuring powder after scooping remove the excess powder to give an accurate volume.
  • Made from food grade PS.
  • Conforms to the latest FDA and EU requirements.
  • BSE/TSE free.
Specifications
Sterility Non-sterile
Capacity (Metric) 100 mL
Disposable Single-use
Width (Metric) 59 mm
Material Polystyrene
Length (Metric) 200 mm