Learn More
Sampling Systems™ SteriWare™ Scraper
Designed to be ergonomic, which means they are easy to use and ideal for scraping and cleaning. Sampling Systems™ SteriWare™ Scraper is ideal for use with foodstuffs, cosmetics and pharmaceuticals.
Brand:  Sampling Systems™ 8785H-75S
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 15164966
  • 2495.00 NOK / Pack of 100
Estimated Shipment: 14-06-2024
Description
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
  • BSE/TSE free
  • Available in 2 sizes
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • No cross contaminaton
  • No cleaning required
Specifications
Specifications
Disposable Scraper
Individually bagged
Natural
75 mm
HDPE
Sterile
190 mm