Learn More
Sampling Systems™ SteriWare™ Sample Kit
Product Code. 15899511 Shop All Sampling Systems Products
Change view
Click to view available options
Capacity (Metric):
30 mL scoop
Unit Size:
Pack of 10
Product Code. Capacity (Metric) unitSize
15899511 30 mL scoop Pack of 10
1 options
Product Code. 15899511
Brand: Sampling Systems™ 8480A30S
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Sampling Systems™ SteriWare™ Sample Kit contains everything you need to take powder samples.
  • Scoops are moulded and packed in a Class 100,000 Cleanroom
  • The Scoop is made from FDA and EU acceptable PS
  • Sterile Sample Bag has volume of 900mL
  • Full batch traceability
  • Pre-sterilized (SAL 10-6)
  • Sterile Sample Bags have a large writing area for recording the details of the sampling activity
Specifications
Includes Scoop and Sample Bag
Capacity (Metric) 30 mL scoop