Learn More
Sampling Systems™ SteriWare™ Open V Spatulas
Improve your quality control by taking fixed volume powder samples easily with SteriWare™ Open V Spatulas. The open tip design is ideal for dispensing and sampling small quantities of powders. Available sterile in a wide range of sizes ideal for the pharmaceutical, chemical and food industry.
Brand:  Sampling Systems™ 8440N-100
Additional Details : Weight : 1.44000kg
 View more versions of this product
Product Code. 17524676
  • 1340.00 NOK / Pack of 100
Estimated Shipment: 18-06-2024
Description
  • Our open ended disposable V spatula are ideal for sampling, raw material testing and general goods in testing.
  • Manufactured to the highest standard in a medical cleanroom to ensure cleanliness and supplied with full documentation.
  • Available in 2 widths and 3 lengths to ensure you have the right sampling tool for the job.
  • The narrow versions are ideal if you want to put the sample into the test tube or take a small sample.
  • The longer version is ideal when you want to take more of a cross sectional sample.
  • The disposable V spatula comes pre bagged and ready to use.
  • Simply unwrap and use.
  • No cleaning or cleaning validation is required.
  • Available in 2 versions, either sterile or non sterile versions.
  • Sterile and non sterile spatulas are ideal when a robust tool is required.
  • Made from high quality PS that conforms to the latest food grade, EC and FDA standards.
  • BSE/TSE free.
  • Batch certificates of conformity supplied.
  • Moulded and packed in a medical cleanroom to maintain cleanliness.
Specifications
Specifications
Type 1 Spatulas
Narrow
White
100 Pk.
10 mm
Polypropylene
Non-sterile
Box
100 mm