Learn More
Sampling Systems™ SteriWare Multi-grip Tweezers
Product Code. 17922122 Shop All Sampling Systems Products
Click to view available options
Length (Metric):
203 mm
300 mm
303 mm
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 50
Product Code. 17922122
Brand: Sampling Systems™ 8270A200S
Only null left
Add to basket
Add to basket
SteriWare multi-grip tweezers can be used as normal tweezers, but also have an additional grip feature enabling the easy and secure handling of small vials and containers.
  • Made from food grade polystyrene
  • Individually bagged to prevent cross contamination
  • 100% manufactured and packed in a cleanroom
  • Available in both sterile and non-sterile
Specifications
Tip Style Serrated, Flat
Length (English) 8 in.
Material Polystyrene
Length (Metric) 203 mm
Product Type Tweezers
Quantity (No. per Case) 50 Tweezers/Cs.
Sterility Sterile
Style Molded