Description
- Assembled and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- Suitable for use in ATEX areas
- Push the sampler into the product, ensuring the sampler is in the closed position
- After leaving the sampler for several seconds to allow powder to enter, rotate handle to close the sampler
- Deposit the sample into a suitable container
- No cross contaminatin
- No cleaning required
Specifications
Specifications
| Product Type | Slot Sampler |
| Material | HDPE |
| Includes | 20 per pack |
| Capacity (Metric) | 50 mL |
| Sterility | Non-sterile |
| Diameter (Metric) | 26 mm |
| Length (Metric) | 620 mm |