Description
- These dosing spoons are a cost effective spoon for sampling, decanting and measuring.
- The double ended spoon means both 2.5 and 5 mL can be measured and sampled from the same spoon.
- They are ideal for use in the hospitals, laboratory, quality control and production environments.
- They are moulded and individually packed in a cleanroom to ensure cleanliness.
Specifications
Specifications
| Sterility | Sterile |
| Capacity (Metric) | 2.5 mL, 5 mL |
| Material | Polystyrene |