Learn More
Sampling Systems™ SteriWare™ Mini Spoons
BIOPHARMA
Product Code. 17524686 Shop All Sampling Systems Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 130
Product Code. Sterility unitSize
17524686 Non-sterile Pack of 130
17534686 Sterile Pack of 130
2 options
Product Code. 17524686
Brand: Sampling Systems™ 8642A01
Measure small quantities of powders and liquids with SteriWare™ Mini Spoons. Available sterile, dosing spoons have a dual 2.5/5 mL bowl ideal for use in healthcare, hospitals, laboratory, and quality control. Each spoon is individually bagged, made from FDA, and EU grade PS.
  • These dosing spoons are a cost effective spoon for sampling, decanting and measuring.
  • The double ended spoon means both 2.5 and 5 mL can be measured and sampled from the same spoon.
  • They are ideal for use in the hospitals, laboratory, quality control and production environments.
  • They are moulded and individually packed in a cleanroom to ensure cleanliness.
Specifications
Sterility Non-sterile
Capacity (Metric) 2.5 mL, 5 mL
Material Polystyrene