Learn More
Sampling Systems™ SteriWare™ Micro-Spatulas
BIOPHARMA
Product Code. 12927935 Shop All Sampling Systems Products
Change view
Click to view available options
Capacity (Metric):
0.3 mL
0.5 mL
2 mL
Color:
Red
White
Material:
HDPE
Polystyrene
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Product Code. Capacity (Metric) Color Material Sterility unitSize
12927935 2 mL White HDPE Non-sterile Pack of 100
15613964 0.5 mL White Polystyrene Sterile Pack of 100
15691739 0.5 mL Red Polystyrene Sterile Pack of 100
15889391 0.5 mL White Polystyrene Non-sterile Pack of 100
15899391 0.3 mL White Polystyrene Non-sterile Pack of 100
15809401 0.3 mL White Polystyrene Sterile Pack of 100
6 options
Product Code. 12927935
Brand: Sampling Systems™ 8015H170
Only null left
Add to basket
Add to basket
Allows samples to be taken from very small containers such as vials with its narrow sampling blade. Sampling Systems™ SteriWare™ Micro-Spatulas are ideal for handling small quantities of powders and granules.
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming materials
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Different blade widths available.  Ideal for small vials or sample containers
  • Also available in HDPE for extra chemical resistance
Specifications
Sterility Non-sterile
Capacity (Metric) 2 mL
Color White
Disposable Single-use
Width (Metric) 13 mm
Material HDPE
Length (Metric) 170 mm