Learn More
Sampling Systems™ SteriWare™ LiquiDip Duo
BIOPHARMA
Product Code. 15899571 Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
80 mL
225 mL
Length (Metric):
450 mm
1100 mm
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 20
Product Code. 15899571
Brand: Sampling Systems™ 8921H1100
Only null left
Add to basket
Add to basket
Sampling Systems™ SteriWare™ LiquiDip Duo is ideal for taking cross sectional or point samples of liquids.
  • Assembled and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
  • BSE/TSE free
  • Suitable for taking cross sectional and point samples
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Available in 2 different lengths
  • Push the sampler into the product, ensuring the top and bottom are in the open position
  • Push the sampler against the base of the container to close the valve
  • Remove the sampler and pour out the liquid into a suitable container
  • No cross contamination
  • No cleaning required
Specifications
Product Type Liquid Sampler
Material HDPE
Includes Valve at both ends
Capacity (Metric) 225 mL
Sterility Non-sterile
Diameter (Metric) Outer 21 mm
Length (Metric) 1100 mm