Learn More
Sampling Systems™ SteriWare™ LiquiDip Duo
Shop All Sampling Systems Products
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
80 mL
225 mL
Length (Metric):
450 mm
1100 mm
Sterility:
Non-sterile
Sterile
missing translation for 'unitSize'
Pack of 20
Description
- Assembled and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
- BSE/TSE free
- Suitable for taking cross sectional and point samples
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- Available in 2 different lengths
- Push the sampler into the product, ensuring the top and bottom are in the open position
- Push the sampler against the base of the container to close the valve
- Remove the sampler and pour out the liquid into a suitable container
- No cross contamination
- No cleaning required
Spécification
Spécification
| Product Type | Liquid Sampler |
| Material | HDPE |
| Includes | Valve at both ends |
| Capacity (Metric) | 80 mL |
| Sterility | Sterile |
| Diameter (Metric) Outer | 21 mm |
| Length (Metric) | 450 mm |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu