Learn More
Sampling Systems™ SteriWare™ HDPE Disposable Powder Thief
BIOPHARMA
Product Code. p-7138913 Shop All Sampling Systems Products
Change view
Click to view available options
Capacity (Metric):
1 mL
5 mL
10 mL
20 mL
Length (Metric):
500 mm
1000 mm
Unit Size:
Pack of 20
Product Code. Capacity (Metric) Length (Metric) unitSize
11935636 10 mL 500 mm Pack of 20
11935457 20 mL 1000 mm Pack of 20
11955437 1 mL 500 mm Pack of 20
11915636 5 mL 1000 mm Pack of 20
11905447 1 mL 1000 mm Pack of 20
11955636 10 mL 1000 mm Pack of 20
11995626 5 mL 500 mm Pack of 20
7 options
Product Code. 11935636
Brand: Sampling Systems™ 8060H510
Only null left
Add to basket
Add to basket
Sampling Systems™ SteriWare™ HDPE Disposable Powder Thief is ideal for taking representative samples from known locations within the bulk powder.
  • Assembled and packed in a cleanroom
  • Individually Bagged
  • Made from FDA or EU conforming HDPE
  • BSE/TSE Free
  • Full Batch Traceability
  • Suitable for use in ATEX zones 0, 1 and 2
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • No cleaning is required before or after sampling
  • No cleaning validation (this is very expensive to set up and run)
  • Manufactured in a class 100,000 cleanroom environment (check individual sampler descriptions)
  • Peace of mind
  • Suitable for all industries (pharmaceutical, food, chemical, cosmetic, biotechnology etc.)
  • Suitable for use in ATEX Zones 0, 1 and 2
  • Financial
  • Save time and money (it will cost more to clean a reusable sampler than to buy a disposable sampler)
Specifications
Material HDPE
Capacity (Metric) 10 mL
Sterility Non-sterile
Diameter (Metric) 21 mm
Length (Metric) 500 mm