Learn More
Sampling Systems™ SteriWare™ HDPE Disposable PharmaScoop™
BIOPHARMA
Product Code. 17418873 Shop All Sampling Systems Products
Change view
Click to view available options
Capacity (Metric):
30 mL
125 mL
500 mL
1000 mL
Unit Size:
Pack of 100
Pack of 25
Pack of 50
Product Code. Capacity (Metric) unitSize
17418873 1000 mL Pack of 25
17438873 500 mL Pack of 50
12987935 30 mL Pack of 100
15869431 125 mL Pack of 100
4 options
Product Code. 17418873
Brand: Sampling Systems™ 8045H1000
Only null left
Add to basket
Add to basket
Use straight from the bag without the need for costly cleaning. Sampling Systems™ SteriWare™ HDPE Disposable PharmaScoop™ is ideal for both sampling and dispensing powders and granules.
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged in ultra-strong packing with easy to open design
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Large variety of different sizes available
  • Good Manufacturing Practice (GMP) to keep your hands out of the product as much as possible
  • Positioned the ergonomic handle high up on the scoop
  • No Cross Contamination
  • No Cleaning required
Specifications
Sterility Non-sterile
Capacity (Metric) 1000 mL
Color White
Disposable Single-use
Product Type Disposable Scoop
Includes Individually Bagged
Material HDPE