Learn More
Sampling Systems™ SteriWare™ FoodScoops
Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
500 mL
1000 mL
2500 mL
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 15
Pack of 25
Pack of 50
Description
- These high quality food scoops are available sterile and non sterile.
- In order to be easily spotted (if they are left behind in the powder) they are manufactured from a highly visible blue food grade polypropylene in line with HACCP guidelines.
- Food Scoops are manufactured and packed under full cleanroom conditions.
- The scoops can be used straight from the bag.
- The robust, ergonomic handles are positioned high up the body to keep the hand away from the powder.
- The scoops are supplied with full documentation including batch certificate of conformity and irradiation certification for the sterilisation.
- The sterile scoops conform to SAL 10-6.
- Made from blue food grade PP.
- Conforms to the latest FDA and EU 10/2011 requirements.
- BSE/TSE free.
- PP is very chemically resistant.
- Range of sizes from 30 mL to 2500 mL
Specifications
Specifications
| Sterility | Non-sterile |
| Capacity (Metric) | 500 mL |
| Color | Blue |
| Disposable | Single-use |
| Material | Polypropylene |