Description
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- No cross contamination
- No cleaning required
Specifications
Specifications
| Material | Polypropylene |
| Capacity (Metric) | 3 L |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Color | White |
| Dimensions (L x W x H) | 310 x 265 x 55 mm |