Learn More
Sampling Systems™ SteriWare™ Disposable Tray
Product Code. 15829461 Shop All Sampling Systems Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 30
Pack of 40
Product Code. Sterility unitSize
15829461 Sterile Pack of 40
30178906 Non-sterile Pack of 30
17161825 Sterile Pack of 30
3 options
Product Code. 15829461
Brand: Sampling Systems™ 8081P3000S
Only null left
Add to basket
Add to basket
Sampling Systems™ SteriWare™ Disposable Tray is ideal for hygienic applications within hospitals and pharmaceutical factories.
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • No cross contamination
  • No cleaning required
Specifications
Material Polypropylene
Capacity (Metric) 3 L
Disposable Single-use
Sterility Sterile
Color White
Dimensions (L x W x H) 310 x 265 x 55 mm