Learn More
Sampling Systems™ SteriWare™ Disposable Slot Sampler
BIOPHARMA
Product Code. 15809461 Shop All Sampling Systems Products
Change view
Click to view available options
Capacity (Metric):
100 mL
170 mL
500 mL
Length (Metric):
620 mm
970 mm
1000 mm
Diameter (Metric):
25 mm
40 mm
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 20
Product Code. Capacity (Metric) Length (Metric) Diameter (Metric) Sterility unitSize
15809461 500 mL 1000 mm 40 mm Non-sterile Pack of 20
12712467 170 mL 970 mm 25 mm Sterile Pack of 20
15442785 170 mL 970 mm 25 mm Non-sterile Pack of 20
15899451 100 mL 620 mm 25 mm Non-sterile Pack of 20
4 options
Product Code. 15809461
Brand: Sampling Systems™ 8073J1000
Only null left
Add to basket
Add to basket
Designed to take a single composite from the top, middle and bottom of your powder bed. Sampling Systems™ SteriWare™ Disposable Slot Samplers are ideal for taking a multilevel, cross sectional sample from powders and granules.
  • Assembled and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Suitable for use in ATEX areas
  • Push the sampler into the product, ensuring the sampler is in the closed position
  • After leaving the sampler for several seconds to allow powder to enter, rotate handle to close the sampler
  • Deposit the sample into a suitable container
  • No cross contaminatin
  • No cleaning required
Specifications
Product Type Disposable Slot Sampler
Material HDPE
Includes 20 per pack
Capacity (Metric) 500 mL
Sterility Non-sterile
Diameter (Metric) 40 mm
Length (Metric) 1000 mm