Description
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- No cross contamination
- No cleaning required
Specifications
Specifications
| Sterility | Sterile |
| Capacity (Metric) | 3 mL |
| Color | White |
| Disposable | Single-use |
| Product Type | Spoon |
| Material | Polystyrene |
| Length (Metric) | 173 mm |