Learn More
Sampling Systems™ SteriWare™ Disposable Powder Spatula
BIOPHARMA
Product Code. p-7138875 Shop All Sampling Systems Products
Click to view available options
Length (Metric):
230 mm
330 mm
Color:
Blue
Red
White
Material:
HDPE
Polypropylene
Polystyrene
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Product Code. 10762674
Brand: Sampling Systems™ 8095A150
Only null left
Add to basket
Add to basket
Easily sample and dispense quantities of powders. Sampling Systems™ SteriWare™ Disposable Powder Spatula is tough and comfortable to use with its specially designed rounded handle and robust blade.
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Suitable for use in ATEX Zones 0, 1 and 2
  • Available in 2 different sizes and a variety of colors
  • No cross contamination
  • No cleaning required
Specifications
Description 150mm Blade
Sterility Non-sterile
Color White
Product Type Disposable Spatula
Width (Metric) 19 mm
Includes 100 Spatulas
Material Polystyrene
Length (Metric) 230 mm