Description
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- No cross contamination
- No cleaning required
Specifications
Specifications
| Diameter (Metric)Top | 250 mm |
| Diameter (Metric) Stem | 40 mm |
| Material | HDPE |
| Autoclavable | Not Autoclavable |
| Graduated | No |
| Sterility | Sterile |
| Product Type | Filling Funnel (Powders) |