Learn More
Sampling Systems™ SteriWare™ Cleaning Coupons
BIOPHARMA
Product Code. p-7239865 Shop All Sampling Systems Products
Change view
Click to view available options
Material:
HDPE
Nylon
PVC
Polypropylene
Polystyrene
Sterility:
Non-sterile
Sterile
Length (Metric):
50 mm
100 mm
Width (Metric):
50 mm
100 mm
Unit Size:
Pack of 50
Product Code. Material Sterility Length (Metric) Width (Metric) unitSize
17594636 HDPE Non-sterile 50 mm 50 mm Pack of 50
17504646 HDPE Sterile 50 mm 50 mm Pack of 50
17534646 Nylon Non-sterile 50 mm 50 mm Pack of 50
17544646 Nylon Sterile 50 mm 50 mm Pack of 50
17574646 Polypropylene Non-sterile 50 mm 50 mm Pack of 50
17584646 Polypropylene Sterile 50 mm 50 mm Pack of 50
17514656 Polystyrene Non-sterile 50 mm 50 mm Pack of 50
17524656 Polystyrene Sterile 50 mm 50 mm Pack of 50
17554656 PVC Non-sterile 50 mm 50 mm Pack of 50
17564656 PVC Sterile 50 mm 50 mm Pack of 50
17574636 HDPE Non-sterile 100 mm 100 mm Pack of 50
17584636 HDPE Sterile 100 mm 100 mm Pack of 50
17514646 Nylon Non-sterile 100 mm 100 mm Pack of 50
17524646 Nylon Sterile 100 mm 100 mm Pack of 50
17554646 Polypropylene Non-sterile 100 mm 100 mm Pack of 50
17564646 Polypropylene Sterile 100 mm 100 mm Pack of 50
17594646 Polystyrene Non-sterile 100 mm 100 mm Pack of 50
17504656 Polystyrene Sterile 100 mm 100 mm Pack of 50
17534656 PVC Non-sterile 100 mm 100 mm Pack of 50
17544656 PVC Sterile 100 mm 100 mm Pack of 50
20 options
Product Code. 17594636
Brand: Sampling Systems™ 8352H50
Only null left
Add to basket
Add to basket
SteriWare™ Cleaning Coupons are essential equipment for checking your cleaning processes. Cleaning coupons are available in a range of different materials and two standard sizes.
  • The SteriWare™ Cleaning coupons are the perfect way to test and validate your cleaning regime.
  • The cleanroom production improves the purity of the final product.
  • Each coupon is 100% inspected and then individually bagged within the cleanroom.
  • Sterile coupons are also available from stock.
  • The coupons are available from stock in the popular sizes, materials and finishes.
  • Supplied with Certificate of Conformity for validation purposes.
  • Popular cleaning coupons include PP, nylon, PS, PVC and HDPE.
  • There are a wide range of other coupons available.
  • Made from high quality virgin material.
  • Individually bagged within the class 10,000 cleanroom.
  • Available with batch specific certificate of conformity.
  • Sterile versions are available with an irradiation certificate.
Specifications
Material HDPE
Length (Metric) 50 mm
Packaging Type Individually Bagged
Sterility Non-sterile
Width (Metric) 50 mm