Learn More
Sampling Systems™ SteriWare™ Cleaning Coupons
BIOPHARMA
Product Code. 17584636 Shop All Sampling Systems Products
Change view
Click to view available options
Material:
HDPE
Nylon
PVC
Polypropylene
Polystyrene
Sterility:
Non-sterile
Sterile
Length (Metric):
50 mm
100 mm
Width (Metric):
50 mm
100 mm
Unit Size:
Pack of 50
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Sterility Length (Metric) Width (Metric) unitSize
17584636 HDPE Sterile 100 mm 100 mm Pack of 50
17594636 HDPE Non-sterile 50 mm 50 mm Pack of 50
17504646 HDPE Sterile 50 mm 50 mm Pack of 50
17534646 Nylon Non-sterile 50 mm 50 mm Pack of 50
17544646 Nylon Sterile 50 mm 50 mm Pack of 50
17574646 Polypropylene Non-sterile 50 mm 50 mm Pack of 50
17584646 Polypropylene Sterile 50 mm 50 mm Pack of 50
17514656 Polystyrene Non-sterile 50 mm 50 mm Pack of 50
17524656 Polystyrene Sterile 50 mm 50 mm Pack of 50
17554656 PVC Non-sterile 50 mm 50 mm Pack of 50
17564656 PVC Sterile 50 mm 50 mm Pack of 50
17574636 HDPE Non-sterile 100 mm 100 mm Pack of 50
17514646 Nylon Non-sterile 100 mm 100 mm Pack of 50
17524646 Nylon Sterile 100 mm 100 mm Pack of 50
17554646 Polypropylene Non-sterile 100 mm 100 mm Pack of 50
17564646 Polypropylene Sterile 100 mm 100 mm Pack of 50
17594646 Polystyrene Non-sterile 100 mm 100 mm Pack of 50
17504656 Polystyrene Sterile 100 mm 100 mm Pack of 50
17534656 PVC Non-sterile 100 mm 100 mm Pack of 50
17544656 PVC Sterile 100 mm 100 mm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Product Code. 17584636 Supplier Sampling Systems™ Supplier No. 8352H100S
SteriWare™ Cleaning Coupons are essential equipment for checking your cleaning processes. Cleaning coupons are available in a range of different materials and two standard sizes.
  • The SteriWare™ Cleaning coupons are the perfect way to test and validate your cleaning regime.
  • The cleanroom production improves the purity of the final product.
  • Each coupon is 100% inspected and then individually bagged within the cleanroom.
  • Sterile coupons are also available from stock.
  • The coupons are available from stock in the popular sizes, materials and finishes.
  • Supplied with Certificate of Conformity for validation purposes.
  • Popular cleaning coupons include PP, nylon, PS, PVC and HDPE.
  • There are a wide range of other coupons available.
  • Made from high quality virgin material.
  • Individually bagged within the class 10,000 cleanroom.
  • Available with batch specific certificate of conformity.
  • Sterile versions are available with an irradiation certificate.
Specifications
Material HDPE
Length (Metric) 100 mm
Packaging Type Individually Bagged
Sterility Sterile
Width (Metric) 100 mm