Learn More
- Home
- Products
- Facility Safety and Maintenance
- Cleaning Supplies and Equipment
- Cleaners and Disinfectants
- Laboratory Cleaning Validation Products
- Sampling Systems™ SteriWare™ Cleaning Coupons
Sampling Systems™ SteriWare™ Cleaning Coupons
Product Code. 17504646
Shop All Sampling Systems Products
Change view
Click to view available options
Material:
HDPE
Nylon
PVC
Polypropylene
Polystyrene
Sterility:
Non-sterile
Sterile
Length (Metric):
50 mm
100 mm
Width (Metric):
50 mm
100 mm
Unit Size:
Pack of 50
20 product options available for selection
| Product Code. | Material | Sterility | Length (Metric) | Width (Metric) | unitSize |
|---|---|---|---|---|---|
| 17504646 | HDPE | Sterile | 50 mm | 50 mm | Pack of 50 |
| 17594636 | HDPE | Non-sterile | 50 mm | 50 mm | Pack of 50 |
| 17534646 | Nylon | Non-sterile | 50 mm | 50 mm | Pack of 50 |
| 17544646 | Nylon | Sterile | 50 mm | 50 mm | Pack of 50 |
| 17574646 | Polypropylene | Non-sterile | 50 mm | 50 mm | Pack of 50 |
| 17584646 | Polypropylene | Sterile | 50 mm | 50 mm | Pack of 50 |
| 17514656 | Polystyrene | Non-sterile | 50 mm | 50 mm | Pack of 50 |
| 17524656 | Polystyrene | Sterile | 50 mm | 50 mm | Pack of 50 |
| 17554656 | PVC | Non-sterile | 50 mm | 50 mm | Pack of 50 |
| 17564656 | PVC | Sterile | 50 mm | 50 mm | Pack of 50 |
| 17574636 | HDPE | Non-sterile | 100 mm | 100 mm | Pack of 50 |
| 17584636 | HDPE | Sterile | 100 mm | 100 mm | Pack of 50 |
| 17514646 | Nylon | Non-sterile | 100 mm | 100 mm | Pack of 50 |
| 17524646 | Nylon | Sterile | 100 mm | 100 mm | Pack of 50 |
| 17554646 | Polypropylene | Non-sterile | 100 mm | 100 mm | Pack of 50 |
| 17564646 | Polypropylene | Sterile | 100 mm | 100 mm | Pack of 50 |
| 17594646 | Polystyrene | Non-sterile | 100 mm | 100 mm | Pack of 50 |
| 17504656 | Polystyrene | Sterile | 100 mm | 100 mm | Pack of 50 |
| 17534656 | PVC | Non-sterile | 100 mm | 100 mm | Pack of 50 |
| 17544656 | PVC | Sterile | 100 mm | 100 mm | Pack of 50 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
Denna artikel kan inte returneras.
Se returpolicy
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
- The SteriWare™ Cleaning coupons are the perfect way to test and validate your cleaning regime.
- The cleanroom production improves the purity of the final product.
- Each coupon is 100% inspected and then individually bagged within the cleanroom.
- Sterile coupons are also available from stock.
- The coupons are available from stock in the popular sizes, materials and finishes.
- Supplied with Certificate of Conformity for validation purposes.
- Popular cleaning coupons include PP, nylon, PS, PVC and HDPE.
- There are a wide range of other coupons available.
- Made from high quality virgin material.
- Individually bagged within the class 10,000 cleanroom.
- Available with batch specific certificate of conformity.
- Sterile versions are available with an irradiation certificate.
Specifikationer
Specifikationer
| Material | HDPE |
| Length (Metric) | 50 mm |
| Packaging Type | Individually Bagged |
| Sterility | Sterile |
| Width (Metric) | 50 mm |
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?