Learn More
Buerkle™ SteriPlast™/LaboPlast™ Bio Sampling Spoon
Shop All Bürkle Products
Click to view available options
Capacity (Metric):
5 mL
20 mL
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 10
Description
- Product made from renewable raw materials
- Bio-PE, white
- Individually packaged and optionally sterilised
- Handle length 300 mm
- For single use
- Clean room manufactured (clean room class 7)
- Complies with EU food requirements and FDA regulations
Specifications
Specifications
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| Capacity (Metric) | 20 mL |
| Certifications/Compliance | Complies with EU food requirements and FDA regulations |
| Disposable | Single-use |
| Product Type | Sampling Spoon |
| Includes | Long Handle |
| Material | Bio-PE |