Learn More
Buerkle™ SteriPlast™ Blue Detectable Scoops
BIOPHARMAGREENER_CHOICE
Product Code. p-7221286 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
50 mL
100 mL
Unit Size:
Pack of 10
Product Code. 17270604
Brand: Buerkle™ 53785016
Only null left
Add to basket
Add to basket
Can be used to take samples of powders, granulates and pastes. The long handle makes it possible to sample larger containers even with little content or to reach through narrow openings.
  • With the straight bottom of the scoop, the medium can be scooped up directly from the bottom of the container
  • Individually packaged and optionally sterilised
  • Handle length 300 mm
  • For single use
  • Clean room manufactured (clean room class 7)
  • Complies with EU food requirements and FDA regulations
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10
Sterility Sterile
Autoclavable Not Autoclavable
Capacity (Metric) 50 mL
Color Blue
Certifications/Compliance Complies with EU food rrequirements and FDA regulations
Disposable Single-use
Product Type Shovel
Length (Metric) Handle 300 mm
Material Polystyrene
Handle Type Long-handled