Learn More
Buerkle™ SteriPlast™ Blue Detectable Sampling Spoon Curved
BIOPHARMAGREENER_CHOICE
Product Code. 17250604 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
10 mL
Unit Size:
Pack of 10
Product Code. 17250604
Brand: Buerkle™ 53785012
Only null left
Add to basket
Add to basket
Suitable for a wide range of different media including powders, granulates, pastes and liquids. The angled spoon shape is also good for reaching into the corners of a container.
  • The long handle of the plastic spoon makes it possible to empty residue from large containers easily and can also reach into places that are hard to reach such as jerrycans with narrow necks.
  • PS, detectable blue
  • Individually packaged and sterilised
  • Can be detected by colour scanners, metal detectors or X-rays
  • Can be used as part of HACCP/IFS/BRC foreign object management
  • Especially for the food, animal feed and pharmaceutical industries
  • Handle length 300 mm
  • For single use
  • Clean room manufactured (clean room class 7)
  • Complies with EU food requirements and FDA regulations
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10
Sterility Sterile
Autoclavable Not Autoclavable
Shape Curved
Capacity (Metric) 10 mL
Color Blue
Certifications/Compliance Complies with EU food rrequirements and FDA regulations
Disposable Single-use
Product Type Detectable Sampling Spoon
Length (Metric) Handle 300 mm
Material Polystyrene
Handle Type Long-handled