Description
- Product and packaging made of renewable raw materials
- Green PE, white
- Individually packaged and sterilised by gamma rays
- Production and packaging according to cleanroom class 7
- EU foodstuffs and FDA approval
Specifications
Specifications
| Sterility | Sterile |
| Color | White |
| Product Type | Spatula |
| Material | Green Polyethylene (Made From Renewable Raw Materials) |
| Length (Metric) | 263 mm |