Learn More
DWK Life Sciences STERIPLAN™ Petri Dish, from Soda-lime Glass
Product Code. p-7189478 Shop All DWK Life Sciences Products
Click to view available options
Diameter (Metric):
40 mm
60 mm
80 mm
90 mm
100 mm
120 mm
150 mm
180 mm
200 mm
Height (Metric):
10 mm
12 mm
15 mm
20 mm
25 mm
30 mm
45 mm
Unit Size:
Pack of 10
Product Code. 11740834
Brand: DWK Life Sciences 237554008
Only null left
Add to basket
Add to basket
Petri dishes are used for the cultivation of bacteria, yeasts, plant and animal cells on solid media.
Cultivation of microorganisms can take place on a gel-like nutrient medium, which is placed in a flat, round, transparent dish with lid (the so-called Petri dish). Due to the solid gel, mixing of the microorganisms is prevented and larger cell culture accumulations (colonies) can grow.
  • Free from bubbles and streaks
  • Even distribution of sewing bottom possible
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
Material Soda Lime Glass
For Use With (Application) Growth and cultivation of microorganisms; Cultivation on solid gel culture medium; Microscopy of microorganisms; Cell culture
No. per Pack 10/Pk.
Height (Metric) 15 mm
Diameter (Metric) 60 mm