Description
Cultivation of microorganisms can take place on a gel-like nutrient medium, which is placed in a flat, round, transparent dish with lid (the so-called Petri dish). Due to the solid gel, mixing of the microorganisms is prevented and larger cell culture accumulations (colonies) can grow.
- Free from bubbles and streaks
- Even distribution of sewing bottom possible
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Material | Soda Lime Glass |
| For Use With (Application) | Growth and cultivation of microorganisms; Cultivation on solid gel culture medium; Microscopy of microorganisms; Cell culture |
| No. per Pack | 10/Pk. |
| Height (Metric) | 15 mm |
| Diameter (Metric) | 90 mm |