Learn More
Sterillin™ Silicone Discs
Click to view available options
Diameter (Metric):
14 mm
40.4 mm
Color:
Black
Red
Unit Size:
Pack of 100
Specifications
Specifications
| Color | Black |
| For Use With (Application) | Used as highly effective cap liners or as a septa for injection ports/pierceable closures and wide variety of applications |
| Material | Rubber |
| Product Type | Disc |
| Diameter (Metric) | 40.4 mm |