Learn More
- Home
- Products
- Dishes
- Weighing Dishes and Papers
- Thermo Scientific™ Sterilin™ Weighing Boats
Thermo Scientific™ Sterilin™ Weighing Boats
Product Code. 11854712
Change view
Click to view available options
Capacity (Metric):
5 mL
7 mL
30 mL
100 mL
250 mL
Color:
Black
White
Type:
Antistatic Weighing Dish
Standard
Weighing Boat
Shape:
Diamond
Square
Unit Size:
Pack of 500
| Product Code. | Capacity (Metric) | Color | Type | Shape | unitSize |
|---|---|---|---|---|---|
| 11854712 | 250 mL | Black | Antistatic Weighing Dish | Square | Pack of 500 |
| 11864722 | 5 mL | Black | Antistatic Weighing Dish | Diamond | Pack of 500 |
| 11864712 | 5 mL | White | Antistatic Weighing Dish | Diamond | Pack of 500 |
| 12427702 | 5 mL | White | Standard | Diamond | Pack of 500 |
| 11894712 | 7 mL | White | Weighing Boat | Square | Pack of 500 |
| 11844712 | 7 mL | Black | Antistatic Weighing Dish | Square | Pack of 500 |
| 11333814 | 7 mL | White | Standard | Square | Pack of 500 |
| 12407702 | 7 mL | Black | Standard | Square | Pack of 500 |
| 11894702 | 250 mL | Black | Standard | Square | Pack of 500 |
| 11804712 | 30 mL | White | Standard | Diamond | Pack of 500 |
| 11824722 | 30 mL | Black | Standard | Diamond | Pack of 500 |
| 11874712 | 30 mL | White | Antistatic Weighing Dish | Diamond | Pack of 500 |
| 11844722 | 30 mL | Black | Antistatic Weighing Dish | Diamond | Pack of 500 |
| 12437702 | 100 mL | White | Standard | Diamond | Pack of 500 |
| 11834722 | 100 mL | Black | Standard | Diamond | Pack of 500 |
| 12447702 | 100 mL | Black | Antistatic Weighing Dish | Square | Pack of 500 |
| 11884712 | 100 mL | White | Antistatic Weighing Dish | Diamond | Pack of 500 |
| 11804722 | 100 mL | White | Antistatic Weighing Dish | Square | Pack of 500 |
| 12417702 | 100 mL | Black | Standard | Square | Pack of 500 |
| 11854722 | 100 mL | Black | Antistatic Weighing Dish | Diamond | Pack of 500 |
| 11824712 | 100 mL | White | Standard | Square | Pack of 500 |
| 11814722 | 250 mL | White | Antistatic Weighing Dish | Square | Pack of 500 |
| 11834712 | 250 mL | White | Standard | Square | Pack of 500 |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
Form a funnel for easy pouring and complete sample transfer using the new and improved Thermo Scientific™ Sterilin™ Weighing Boats, featuring radiused corners. All sizes packed 250 per pack, 500 per case.
- Manufactured from high impact polystyrene
- Hydrophobic surface ensures no absorption of water from atmosphere or sample
- Available in black or white to ensure suitability for different colored sample materials
- Anti-static range alleviates issues with powder hang-up
- Flat base for a secure, firm rest on the balance pan
- Supplied nonsterile
Specifications
Specifications
| Bottom Shape | Flat |
| Capacity (Metric) | 250 mL |
| Color | Black |
| Material | Polystyrene |
| Shape | Square |
| Sterility | Non-sterile |
| Type | Antistatic Weighing Dish |
| Length (Metric) | 140 mm |
| Width (Metric) | 140 mm |