Learn More
Thermo Scientific™ Sterilin™ Weighing Boats
Product Code. 11824712
Change view
Click to view available options
Capacity (Metric):
5 mL
7 mL
30 mL
100 mL
250 mL
Color:
Black
White
Type:
Antistatic Weighing Dish
Standard
Weighing Boat
Shape:
Diamond
Square
Unit Size:
Pack of 500
Product Code. Capacity (Metric) Color Type Shape unitSize
11824712 100 mL White Standard Square Pack of 500
11864722 5 mL Black Antistatic Weighing Dish Diamond Pack of 500
11864712 5 mL White Antistatic Weighing Dish Diamond Pack of 500
12427702 5 mL White Standard Diamond Pack of 500
11894712 7 mL White Weighing Boat Square Pack of 500
11844712 7 mL Black Antistatic Weighing Dish Square Pack of 500
11333814 7 mL White Standard Square Pack of 500
12407702 7 mL Black Standard Square Pack of 500
11894702 250 mL Black Standard Square Pack of 500
11804712 30 mL White Standard Diamond Pack of 500
11824722 30 mL Black Standard Diamond Pack of 500
11874712 30 mL White Antistatic Weighing Dish Diamond Pack of 500
11844722 30 mL Black Antistatic Weighing Dish Diamond Pack of 500
12437702 100 mL White Standard Diamond Pack of 500
11834722 100 mL Black Standard Diamond Pack of 500
12447702 100 mL Black Antistatic Weighing Dish Square Pack of 500
11884712 100 mL White Antistatic Weighing Dish Diamond Pack of 500
11804722 100 mL White Antistatic Weighing Dish Square Pack of 500
12417702 100 mL Black Standard Square Pack of 500
11854722 100 mL Black Antistatic Weighing Dish Diamond Pack of 500
11854712 250 mL Black Antistatic Weighing Dish Square Pack of 500
11814722 250 mL White Antistatic Weighing Dish Square Pack of 500
11834712 250 mL White Standard Square Pack of 500
23 options
Product Code. 11824712
Brand: Thermo Scientific™ WB30254
Form a funnel for easy pouring and complete sample transfer using the new and improved Thermo Scientific™ Sterilin™ Weighing Boats, featuring radiused corners.
Form a funnel for easy pouring and complete sample transfer using the new and improved Thermo Scientific™ Sterilin™ Weighing Boats, featuring radiused corners. All sizes packed 250 per pack, 500 per case.
  • Manufactured from high impact polystyrene
  • Hydrophobic surface ensures no absorption of water from atmosphere or sample
  • Available in black or white to ensure suitability for different colored sample materials
  • Anti-static range alleviates issues with powder hang-up
  • Flat base for a secure, firm rest on the balance pan
  • Supplied nonsterile
TRUSTED_SUSTAINABILITY
Specifications
Bottom Shape Flat
Capacity (Metric) 100 mL
Color White
Material Polystyrene
Shape Square
Sterility Non-sterile
Type Standard
Length (Metric) 80 mm
Width (Metric) 80 mm