Learn More
Thermo Scientific™ Sterilin™ Silicone Tubing
Product Code. 15157172
Change view
Click to view available options
Diameter (Metric) Inner:
0.5 mm
0.8 mm
1 mm
1.5 mm
2 mm
2.5 mm
3 mm
3.2 mm
4 mm
4.5 mm
4.7 mm
5 mm
5.5 mm
6 mm
6.3 mm
7.5 mm
8 mm
8.5 mm
9 mm
9.5 mm
10 mm
12.7 mm
16 mm
19 mm
Diameter (Metric) Outer:
1 mm
1.5 mm
2 mm
2.5 mm
3.5 mm
3.7 mm
4.1 mm
4.2 mm
4.5 mm
4.8 mm
5 mm
5.2 mm
6 mm
6.4 mm
7 mm
7.0 mm
7.2 mm
7.7 mm
7.95 mm
8.0 mm
8.2 mm
9.5 mm
10 mm
10.5 mm
11.2 mm
11.3 mm
12 mm
12.7 mm
14 mm
14.4 mm
15.4 mm
15.9 mm
16 mm
16.5 mm
17 mm
18 mm
19.1 mm
22.4 mm
25.4 mm
Thickness (Metric):
0.25 mm
0.5 mm
0.8 mm
1 mm
1.2 mm
1.5 mm
1.6 mm
1.7 mm
1.75 mm
2 mm
2.0 mm
2.5 mm
3 mm
3.2 mm
4 mm
5 mm
Unit Size:
15m
Pack of 15
Product Code. Diameter (Metric) Inner Diameter (Metric) Outer Thickness (Metric) unitSize
15157172 0.5 mm 1.5 mm 0.5 mm 15m
13180423 0.5 mm 1 mm 0.25 mm Pack of 15
12802983 0.5 mm 3.7 mm 1.6 mm 15m
12892973 0.8 mm 4.2 mm 1.7 mm 15m
12812983 1 mm 2 mm 0.5 mm 15m
12832983 1 mm 5 mm 2 mm 15m
12842983 1.5 mm 2.5 mm 0.5 mm 15m
12852983 1.5 mm 3.5 mm 1 mm 15m
12862983 1.5 mm 4.5 mm 1.5 mm 15m
12892983 2 mm 4.5 mm 1.2 mm 15m
12802993 2.5 mm 4.1 mm 0.8 mm 15m
12812993 2.5 mm 4.5 mm 1 mm 15m
13199215 3 mm 7.0 mm 2.0 mm 15m
12832993 3.2 mm 4.8 mm 0.8 mm 15m
12852993 3.2 mm 5.2 mm 1 mm 15m
12842993 3.2 mm 6.4 mm 1.6 mm 15m
12862993 4 mm 6 mm 1 mm 15m
12882993 4 mm 7.2 mm 1.6 mm 15m
12892993 4 mm 8.0 mm 2.0 mm 15m
12803003 4.5 mm 7.7 mm 1.6 mm 15m
12813003 4.7 mm 7.95 mm 1.6 mm 15m
12823003 5 mm 7 mm 1 mm 15m
12833003 5 mm 8.2 mm 1.6 mm 15m
12853003 5.5 mm 10.5 mm 2.5 mm 15m
12863003 6 mm 9.5 mm 1.75 mm 15m
12873003 6 mm 10 mm 2 mm 15m
12883003 6 mm 14 mm 4 mm 15m
12893003 6.3 mm 9.5 mm 1.6 mm 15m
13111454 6.3 mm 11.3 mm 2.5 mm 15m
13190423 6.3 mm 12.7 mm 3.2 mm 15m
12833013 7.5 mm 10.5 mm 1.5 mm 15m
12843013 8 mm 11.2 mm 1.6 mm 15m
12853013 8 mm 12 mm 2 mm 15m
13121454 8 mm 14.4 mm 3.2 mm 15m
13100433 8 mm 16 mm 4 mm 15m
12893013 8 mm 18 mm 5 mm 15m
12803023 8.5 mm 16.5 mm 4 mm 15m
12813023 9 mm 14 mm 2.5 mm 15m
12823023 9 mm 15.4 mm 3.2 mm 15m
12833023 9 mm 17 mm 4 mm 15m
12843023 9.5 mm 15.9 mm 3.2 mm 15m
12853023 10 mm 14 mm 2 mm 15m
12863023 10 mm 16 mm 3 mm 15m
13131454 12.7 mm 19.1 mm 3.2 mm 15m
12883023 16 mm 22.4 mm 3.2 mm 15m
12893023 19 mm 25.4 mm 3.2 mm 15m
46 options
Product Code. 15157172
Brand: Thermo Scientific™ TSR0050050P
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Perform various applications with high quality, silicone tubing made from nontoxic material.
Perform various applications with high-quality silicone tubing made from nontoxic material.
  • Laser etched batch code for ease of traceability
  • Ideal for use with peristaltic pumps
  • Can be autoclaved or dry heat sterilized
  • 15m length
  • Manufactured in ISO Class 7 clean room
TRUSTED_SUSTAINABILITY
Specifications
Description Silicone tubing
Autoclavable Autoclavable
Diameter (Metric) Inner 0.5 mm
Material Silicone
Diameter (Metric) Outer 1.5 mm
Overall Length (English) 49 ft.
Overall Length (Metric) 15 m
Thickness (Metric) 0.5 mm
Color Translucent
Flexibility Very Flexible
Product Type Flexible Tubing