Learn More
- Home
- Products
- Tubing
- Plastic Tubing
- Thermo Scientific™ Sterilin™ Silicone Tubing
Thermo Scientific™ Sterilin™ Silicone Tubing
Product Code. 12892993| Codice prodotto. | Diameter (Metric) Inner | Diameter (Metric) Outer | Thickness (Metric) | unitSize |
|---|---|---|---|---|
| 12892993 | 4 mm | 8.0 mm | 2.0 mm | 15m |
| 13180423 | 0.5 mm | 1 mm | 0.25 mm | Pack of 15 |
| 15157172 | 0.5 mm | 1.5 mm | 0.5 mm | 15m |
| 12802983 | 0.5 mm | 3.7 mm | 1.6 mm | 15m |
| 12892973 | 0.8 mm | 4.2 mm | 1.7 mm | 15m |
| 12812983 | 1 mm | 2 mm | 0.5 mm | 15m |
| 12832983 | 1 mm | 5 mm | 2 mm | 15m |
| 12842983 | 1.5 mm | 2.5 mm | 0.5 mm | 15m |
| 12852983 | 1.5 mm | 3.5 mm | 1 mm | 15m |
| 12862983 | 1.5 mm | 4.5 mm | 1.5 mm | 15m |
| 12892983 | 2 mm | 4.5 mm | 1.2 mm | 15m |
| 12802993 | 2.5 mm | 4.1 mm | 0.8 mm | 15m |
| 12812993 | 2.5 mm | 4.5 mm | 1 mm | 15m |
| 13199215 | 3 mm | 7.0 mm | 2.0 mm | 15m |
| 12832993 | 3.2 mm | 4.8 mm | 0.8 mm | 15m |
| 12852993 | 3.2 mm | 5.2 mm | 1 mm | 15m |
| 12842993 | 3.2 mm | 6.4 mm | 1.6 mm | 15m |
| 12862993 | 4 mm | 6 mm | 1 mm | 15m |
| 12882993 | 4 mm | 7.2 mm | 1.6 mm | 15m |
| 12803003 | 4.5 mm | 7.7 mm | 1.6 mm | 15m |
| 12813003 | 4.7 mm | 7.95 mm | 1.6 mm | 15m |
| 12823003 | 5 mm | 7 mm | 1 mm | 15m |
| 12833003 | 5 mm | 8.2 mm | 1.6 mm | 15m |
| 12853003 | 5.5 mm | 10.5 mm | 2.5 mm | 15m |
| 12863003 | 6 mm | 9.5 mm | 1.75 mm | 15m |
| 12873003 | 6 mm | 10 mm | 2 mm | 15m |
| 12883003 | 6 mm | 14 mm | 4 mm | 15m |
| 12893003 | 6.3 mm | 9.5 mm | 1.6 mm | 15m |
| 13111454 | 6.3 mm | 11.3 mm | 2.5 mm | 15m |
| 13190423 | 6.3 mm | 12.7 mm | 3.2 mm | 15m |
| 12833013 | 7.5 mm | 10.5 mm | 1.5 mm | 15m |
| 12843013 | 8 mm | 11.2 mm | 1.6 mm | 15m |
| 12853013 | 8 mm | 12 mm | 2 mm | 15m |
| 13121454 | 8 mm | 14.4 mm | 3.2 mm | 15m |
| 13100433 | 8 mm | 16 mm | 4 mm | 15m |
| 12893013 | 8 mm | 18 mm | 5 mm | 15m |
| 12803023 | 8.5 mm | 16.5 mm | 4 mm | 15m |
| 12813023 | 9 mm | 14 mm | 2.5 mm | 15m |
| 12823023 | 9 mm | 15.4 mm | 3.2 mm | 15m |
| 12833023 | 9 mm | 17 mm | 4 mm | 15m |
| 12843023 | 9.5 mm | 15.9 mm | 3.2 mm | 15m |
| 12853023 | 10 mm | 14 mm | 2 mm | 15m |
| 12863023 | 10 mm | 16 mm | 3 mm | 15m |
| 13131454 | 12.7 mm | 19.1 mm | 3.2 mm | 15m |
| 12883023 | 16 mm | 22.4 mm | 3.2 mm | 15m |
| 12893023 | 19 mm | 25.4 mm | 3.2 mm | 15m |
Codice prodotto. 12892993
Marca: Thermo Scientific™ TSR0400200P
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
Description
Perform various applications with high-quality silicone tubing made from nontoxic material.
- Laser etched batch code for ease of traceability
- Ideal for use with peristaltic pumps
- Can be autoclaved or dry heat sterilized
- 15m length
- Manufactured in ISO Class 7 clean room
Specifications
Specifications
| Description | Silicone tubing |
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 4 mm |
| Material | Silicone |
| Diameter (Metric) Outer | 8.0 mm |
| Overall Length (English) | 49 ft. |
| Overall Length (Metric) | 15 m |
| Thickness (Metric) | 2.0 mm |
| Color | Translucent |
| Flexibility | Very Flexible |
| Product Type | Flexible Tubing |
| Show More Show Less |