Learn More
Corning™ PYREX™ Disposable Round Bottom Threaded Culture Tubes, Without Marking Spot or Caps
Shop All Corning ProductsDescription
A round bottom tube without marking spot. Tubes are made of borosilicate glass and are sold without caps.
For caps, see Cat. No. 99999.
Specifications
Specifications
| Design Type | Screw neck |
| Capacity (Metric) | 11.5 mL |
| Length (Metric) | 100 mm |
| Material | Borosilicate Glass |
| Disposable | Single-use |
| For Use With | Ideal for use with samples sensitive to leaching from plastic tubes |
| Label Type | None |
| Packaging | Bulk |
| Thread Size | 15-415 |
| Sterility | Non-sterile |
| Show More |