Learn More
Thermo Scientific™ Sterilin™ Round Base Rimless Borosilicate Glass Rimless Tubes
GREENER_CHOICE
Product Code. 10613941
Change view
Click to view available options
Capacity (Metric):
10 mL
11 mL
15 mL
19 mL
23 mL
28.5 mL
36 mL
Diameter (Metric):
13 mm
15 mm
16 mm
18 mm
20 mm
Height (Metric):
85 mm
100 mm
125 mm
150 mm
No. per Case:
500
1000
Unit Size:
Pack of 1000
Pack of 500
Product Code. Capacity (Metric) Diameter (Metric) Height (Metric) No. per Case unitSize
10613941 11 mL 15 mm 85 mm 1000 Pack of 1000
11379243 10 mL 13 mm 100 mm 1000 Pack of 1000
11389243 15 mL 16 mm 100 mm 1000 Pack of 1000
11309253 19 mL 16 mm 125 mm 1000 Pack of 1000
11319253 23 mL 16 mm 150 mm 1000 Pack of 1000
11349253 36 mL 20 mm 150 mm 500 Pack of 500
11329253 28.5 mL 18 mm 150 mm 500 Pack of 500
7 options
Product Code. 10613941
Brand: Thermo Scientific™ 9944515
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Trust these round-base borosilicate glass tubes in high temperature, aggressive chemical applications. Ideal for use with samples that are sensitive to plastic tube leachables.
Trust these round-base borosilicate glass tubes in high temperature, aggressive chemical applications. Thermo Scientific™ Sterilin™ Round Base Rimless Borosilicate Glass Tubes are ideal for use with samples that are sensitive to plastic tube leachables.
NOT AVAILABLE IN AMERICA
  • Manufactured from Pyrex borosilicate glass
  • High resistance to temperature and chemical attack
  • Tube profile rimless
  • Supplied without caps
  • Autoclavable at 121°C
Ordering Information
  • Available in UK only
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 15 mm
Product Type Rimless Tube
Autoclavable Autoclavable
Capacity (Metric) 11 mL
Material Borosilicate Glass
No. per Case 1000
No. per Pack 250
Sterility Non-sterile
Packaging Quantity 1000 Case (250 Pack)
Capacity (English) 0.3719 fl oz.
Height (Metric) 85 mm