Learn More
Thermo Scientific™ Sterilin™ Polystyrene Containers, 60mL to 250mL
GREENER_CHOICE
Product Code. p-4525838
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
60 mL
100 mL
150 mL
250 mL
Closure Material:
Metal
Polyethylene
Label:
No Label
Plain
Printed
No. per Case:
120/Cs.
200/Cs.
300/Cs.
50/Cs.
missing translation for 'unitSize'
Pack of 120
Pack of 200
Pack of 300
Pack of 50
Product Code. 11379123
missing translation for 'mfr': Thermo Scientific™ 125AM
Contain samples in Thermo Scientific™ Sterilin™ Polystyrene Containers with 60mL to 250mL capacities. Available with either plastic or metal flow seal caps.
Contain samples in Thermo Scientific™ Sterilin™ Polystyrene Containers with 60mL to 250mL capacities. Available with either plastic or metal flow seal caps.
NOT AVAILABLE IN AMERICA
Features:
  • Aseptically manufactured
  • Leak-tested in accordance with BS EN 14254 Annexe D
Note:
For in vitro use only.
Ordering Information:
Product is not available in North America.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Container
Closure Included Yes
Material Polystyrene
Shape Cylindrical
Sterility Non-sterile
Capacity (Metric) 60 mL
Certifications/Compliance Leak-tested in accordance with BS EN ISO 6717
Closure Material Metal
Diameter (Metric) 44.5 mm
Height (Metric) 61 mm
Label No Label
No. per Case 300/Cs.