Learn More
Thermo Scientific™ Sterilin™ Autoclave Bags
Click to view available options
Dimensions (L x W):
601 x 406mm
610 x 406mm
660 x 305mm
810 x 610mm
Material:
HDPE
Polypropylene
No. per Case:
200
500
Unit Size:
Pack of 200
Pack of 500
Description
Sterilin Autoclave Bags are strong disposable bags with blue biohazard printing specifically designed for contaminated waste disposal in autoclaves or incinerators.
- HDPE bags are suitable for sterilization at 121°C and polypropylene bags are suitable for the inactivation of particularly resistant biological waste at 135°C
- Convenient “tissue box” cartons dispense bags individually
- Coated wire stands or cardboard holders available for use with autoclave bags
Ordering Alerts:
Not available in all geographic regions
Specifications
Specifications
| Description | Autoclave bags (HDPE) |
| Material | HDPE |
| No. per Case | 500 |
| Temperature Range (Metric) | 121°C |
| Dimensions (L x W) | 660 x 305mm |
| Length (English) | 26 in. |
| Length (Metric) | 660 mm |
| Width (English) | 12 in. |
| Width (Metric) | 305 mm |