Learn More
Thermo Scientific™ Sterilin™ Autoclave Bags
Product Code. 11329103
Change view
Click to view available options
Dimensions (L x W):
601 x 406mm
610 x 406mm
660 x 305mm
810 x 610mm
Material:
HDPE
Polypropylene
No. per Case:
200
500
Unit Size:
Pack of 200
Pack of 500
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Material No. per Case Unit Size
11329103 810 x 610mm Polypropylene 200 Pack of 200
11399093 660 x 305mm HDPE 200 Pack of 200
11309103 610 x 406mm HDPE 200 Pack of 200
11359103 660 x 305mm HDPE 500 Pack of 500
11844145 610 x 406mm HDPE 500 Pack of 500
11339103 810 x 610mm HDPE 200 Pack of 200
11379103 660 x 305mm Polypropylene 200 Pack of 200
11349103 660 x 305mm Polypropylene 500 Pack of 500
11389103 610 x 406mm Polypropylene 200 Pack of 200
11369103 601 x 406mm Polypropylene 500 Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 11329103 Supplier Thermo Scientific™ Supplier No. 511HT
  • ONLINE EXCLUSIVE PRICE
    3175.00 NOK
    2988.64 NOK / Pack of 200
    Save 186.36 NOK
    5% Off
Estimated Shipment: 28-12-2026
Sterilin Autoclave Bags are strong disposable bags with blue biohazard printing specifically designed for contaminated waste disposal in autoclaves or incinerators.
  • HDPE bags are suitable for sterilization at 121°C and polypropylene bags are suitable for the inactivation of particularly resistant biological waste at 135°C
  • Convenient “tissue box” cartons dispense bags individually
  • Coated wire stands or cardboard holders available for use with autoclave bags
Ordering Alerts:
Not available in all geographic regions
TRUSTED_SUSTAINABILITY
Specifications
Description Autoclave bags (PP)
Material Polypropylene
No. per Case 200
Temperature Range (Metric) 135°C
Dimensions (L x W) 810 x 610mm
Length (English) 32 in.
Width (English) 24 in.
Length (Metric) 810 mm
Width (Metric) 610 mm