Learn More
Bel-Art™ Sterileware™ Sterile Styrene Sampler Spoons
Product Code. p-166485
Change view
Click to view available options
Capacity (Metric):
1.23 mL
2.46 mL
4.93 mL
14.79 mL
Quantity:
10/Pk.
200/Cs.
Unit Size:
Pack of 10
Pack of 200
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Quantity Unit Size
11761566 14.79 mL 200/Cs. Pack of 200
15500142 1.23 mL 10/Pk. Pack of 10
11378733 2.46 mL 200/Cs. Pack of 200
15530142 14.79 mL 10/Pk. Pack of 10
10573924 1.23 mL 200/Cs. Pack of 200
15510142 2.46 mL 10/Pk. Pack of 10
15520142 4.93 mL 10/Pk. Pack of 10
11316944 4.93 mL 200/Cs. Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11761566 Supplier Bel-Art™ Supplier No. H369460000
Add to basket
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Polystyrene plastic sterile sampler spoons are designed to gather small amounts of sample material.
Manufactured from polystyrene plastic. SP Scienceware™ Sterileware™ Sterile Styrene Sampler Spoons are designed to gather small amounts of sample material.
  • Sterile, individually packaged, sterilized by gamma irradiation, lot stamped for accurate lot tracing, and shipped with a Certificate of Processing for Sterilization
  • FDA grade materials; safe for food, drug and cosmetic contact
  • Long handle reaches easily into jars, bottles and other confined areas, and minimizes the potential for personal contact with sampled material
  • Overall length 17.8 cm (7 in.)
Spezifikation
Quantity 200/Cs.
Capacity (English) 3 tsp.
Capacity (Metric) 14.79 mL
Color White
Certifications/Compliance FDA
Disposable Single-use
Product Type Spoon
For Use With (Application) Designed to gather small amounts of sample material
Length (English) 7 in.
Width (English) 1.53 in.
Width (Metric) 4 cm
Dimensions (L x W) 17.8 x 4 cm
Includes Certificate of sterilization
Material Polystyrene
Handle Type Long
Length (Metric) 17.8 cm
Sterility Sterile
Autoclavable Not Autoclavable
Shape Spoon
Mehr anzeigen Weniger anzeigen