missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Solveco Sterile Spray Trigger
Description
- Sterile trigger spray for easy and effective dispensing.
- The trigger is individually packaged in double bags with an additional outer bag, creating triple-layer packaging, and then sterilized with 25 kGy.
- Each bag is labeled with its batch number, production date, item number, and expiration date.
Specifications
Specifications
| Color | White |
| Material | HDPE |
| Sterility | Sterile |
| Disposable | Single-use |
| Fit for Purpose | Sterile |
| For Use With (Equipment) | 1 L Bottles |
| Autoclavable | Not Autoclavable |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?