Learn More
Gosselin™ Sterile Spoons
Product Code. p-8002796
Change view
Click to view available options
Capacity (Metric):
2 mL
8 mL
Unit Size:
Pack of 100
Pack of 1000
Product Code. Capacity (Metric) unitSize
12304119 8 mL Pack of 1000
11810522 2 mL Pack of 100
2 options
Product Code. 12304119
Brand: Gosselin™ CUIG02
Only null left
Add to basket
Add to basket
Suitable for general laboratory use. Gosselin™ Sterile Spoons are manufactured from sterile polystyrene.
  • Individually packaged
  • Available in two different sizes
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 8 mL
Color White
Certifications/Compliance EC N°1935/2004, ISO 11137