Learn More
Fisherbrand™ Sterile Sample Bags
Product Code. 17943182 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W):
4.5 x 9 in.
12 x 18 in.
15 x 20 in.
Quantity:
100/Pk.
250/Pk.
500/Pk.
Unit Size:
Pack of 100
Pack of 250
Pack of 500
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Quantity unitSize
17943182 4.5 x 9 in. 500/Pk. Pack of 500
17953182 12 x 18 in. 250/Pk. Pack of 250
17903192 15 x 20 in. 100/Pk. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17943182
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
An economical and efficient way to collect, contain and carry samples.
  • These sample bags are ideal for transportation and storage of solids, semisolids, and liquids.
  • Sterile
  • Highly resistant
  • Wire closure for a superior seal
Specifications
Autoclavable Not Autoclavable
Color Clear
Material Polyethylene
Sterility Sterile
Quantity 500/Pk.
Dimensions (L x W) 4.5 x 9 in.
Height (Metric) 229 mm
Length (Metric) 114 mm
Thickness (Metric) 70μm