Learn More
Corning™ Sterile Polystyrene Inoculating Loops
GREENER_CHOICE
Product Code. 11769515
Change view
Click to view available options
Volume:
1 μL
10 μL
Product Type:
Inoculating Loop
Inoculating Loop, Needle Tip
Inoculating Loop, Spherical Tip
Unit Size:
Pack of 3000
Pack of 9000
Product Code. Volume Product Type unitSize
11769515 10 μL Inoculating Loop Pack of 9000
12682275 1 μL Inoculating Loop, Needle Tip Pack of 9000
11733124 1 μL Inoculating Loop, Needle Tip Pack of 3000
15241065 1 μL Inoculating Loop, Spherical Tip Pack of 9000
11763124 10 μL Inoculating Loop, Needle Tip Pack of 3000
5 options
Product Code. 11769515
Brand: Corning™ OS10B02
Only null left
Add to basket
Add to basket
Constructed of flexible polystyrene for optimum retrieval of an inoculum from a culture. Gosselin™ Sterile Polystyrene Inoculating Loops provide a superior solution for transferring microscopic organisms, inoculum, or streaking.
  • Polystyrene (PS) inoculating loops
  • Spherical or needle extremity
  • 1 μl or 10 μl
TRUSTED_SUSTAINABILITY
Specifications
Volume 10 μL
Length (Metric) 195 mm
Material Polystyrene
Sterility Sterile
Color Blue
Product Type Inoculating Loop
Quantity 150 BG