Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Sterile Components Stoppers
Product Code. 16267290 Shop All Wheaton Science Products Products
Change view
Click to view available options
Closure Size:
13 mm
20 mm
Style:
OmniFlex 3G Igloo
OmniFlex 3G Straight Plug
Ultra Pure Straight Plug
Unit Size:
Pack of 220
Pack of 230
Product Code. Closure Size Style unitSize
16267290 20 mm Ultra Pure Straight Plug Pack of 220
16227290 13 mm OmniFlex 3G Straight Plug Pack of 230
16257290 20 mm OmniFlex 3G Straight Plug Pack of 220
16237290 13 mm Ultra Pure Straight Plug Pack of 230
16247290 13 mm OmniFlex 3G Igloo Pack of 230
16277290 20 mm OmniFlex 3G Igloo Pack of 220
6 options
Product Code. 16267290
Brand: DWK Life Sciences W986007S
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
  • Sterile
  • Stopper
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
TRUSTED_SUSTAINABILITY
Specifications
Sterility Sterile
No. per Case 220
Style Ultra Pure Straight Plug
Product Line Wheaton CompletePAK
Closure Size 20 mm
Certifications/Compliance USP 788, 85, and 71, Container Closure Integrity (CCI) certified