Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Sterile Components Stoppers
Product Code. 16237290 Shop All Wheaton Science Products Products
Click to view available options
Closure Size:
13 mm
20 mm
Style:
OmniFlex 3G Igloo
OmniFlex 3G Straight Plug
Ultra Pure Straight Plug
Unit Size:
Pack of 220
Pack of 230
Product Code. 16237290
Brand: DWK Life Sciences W986004S
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
  • Sterile
  • Stopper
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
TRUSTED_SUSTAINABILITY
Specifications
Sterility Sterile
No. per Case 230
Style Ultra Pure Straight Plug
Product Line Wheaton CompletePAK
Closure Size 13 mm
Certifications/Compliance USP 788, 85, and 71, Container Closure Integrity (CCI) certified