Learn More
Technical Service Consultants™ Sterile Inoculation Needle
Description
- Tapered tip for improved bead security
- Safe and secure transfer of cultures from freezer to culture plate
- Eliminates need to balance beads with inoculating loop
Specifications
Specifications
| Disposable | Single-use |
| Material | Polystyrene |
| Sterility | Sterile |
| Color | Clear |
| Product Type | Inoculating Loop |
| For Use With (Equipment) | General laboratory use only |
| Quantity | 1000/Pk. |
| For Use With (Application) | General Laboratory use only |