Learn More
WLD Tec™ Sterile Inoculation Loop
Click to view available options
Diameter (Metric) Inside:
3 mm
5 mm
Unit Size:
Pack of 10
Specifications
Specifications
| Disposable | Single-use |
| Diameter (Metric) Inside | 5 mm |
| Material | Stainless Steel |
| Sterility | Sterile |
| Product Type | Inoculating Loop |
| For Use With (Equipment) | Laboratory and Research use |
| Quantity | 10/pk. |
| For Use With (Application) | Laboratory and Research use |