Description
SteriBag sterilized sampling bags are ideal for samples in chemistry, pharmaceuticals, medicine, veterinary science and foodstuffs. The bags are made of polyethylene and are therefore exceptionally tearproof and leakproof.
- Sterilized
- Can be labelled
- Leakproof
- Save space
- Transparent
- With practical tear-open closure
- Proven millions of times
Specifications
Specifications
| Capacity (Metric) | 650 mL |
| Closure Type | Tear-open |
| Color | Clear |
| For Use With (Application) | Ideal for Samples in Chemistry, Pharmaceuticals, Medicine, Veterinary Science and Foodstuffs |
| Material | Polyethylene |
| Sterility | Sterile |
| Quantity | 500/Pk. |
| Length (Metric) | 229 mm |
| Width (Metric) | 114 mm |
| Product Type | Sample Bag |